Description
- Accessory
- For use with moisture analyzer
Specifications
Specifications
| Width (Metric) | 14 mm |
| Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) |
| Quantity | 3/Pk. |
| Weight (Metric) | 160 g |