missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REV1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | REV1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
REV1 Polyclonal antibody specifically detects REV1 in Human samples. It is validated for Western BlotSpecifications
| REV1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Direct Reversal of DNA Damage | |
| PBS (pH 7.3), 50% glycerol | |
| 51455 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AIBP80, Alpha integrin-binding protein 80, DNA repair protein REV1, EC 2.7.7.-, FLJ21523, MGC163283, MGC26225, REV1 (yeast homolog)- like, REV1 homolog (S. cerevisiae), REV1- like, REV1L, REV1-like (yeast), Rev1-like terminal deoxycytidyl transferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-345 of human REV1 (NP_057400.1). AGRLLSYIPYQLYTKQSSVQKGLSFNPVCRPEDPLPGPSNIAKQLNNRVNHIVKKIETENEVKVNGMNSWNEEDENNDFSFVDLEQTSPGRKQNGIPHPRGSTAIFNGHTPSSNGALKTQDCLVPMVNSVASRLSPAFSQEEDKAEKSSTDFRDCTLQQLQQSTRNTDALRNPHRTNSFSLSPLHSNTKINGAHHSTVQGPSSTKSTSSVSTFSKAAPSVPSKPSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title