Learn More
BUCHI FlashPure ID Silica Flash Chromatography Cartridges
Product Code. 16360276 Shop All Buchi Products
Change view
Click to view available options
Bed Weight (Metric):
80 g
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight (Metric) unitSize
16360276 80 g Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16360276 Supplier BUCHI Supplier No. 145146133
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
20 Pieces
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
TRUSTED_SUSTAINABILITY
Specifications
Material Silica
Type Reveleris
Includes 12 Pc.
Bed Weight (Metric) 80 g
For Use With (Application) Useful for Conventional Applications; Polar Compounds
Max. Pressure 200 psig