missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REXO2 Antibody (59D9), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42639AF750
This item is not returnable.
View return policy
Description
REXO2 Monoclonal specifically detects REXO2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| REXO2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| REXO2 | |
| Recombinant human REXO 2 (26-237aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 59D9 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| CGI-114, DKFZP566E144, EC 3.1, oligoribonuclease, mitochondrial, REX2, REX2, RNA exonuclease 2 homolog (S. cerevisiae), RFN, RNA exonuclease 2 homolog, SFNMGC111570, Small fragment nuclease, SMFN | |
| Mouse | |
| Protein A purified | |
| DNA replication Transcription Translation and Splicing | |
| 25996.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction