missing translation for 'onlineSavingsMsg'
Learn More

REXO2 Monoclonal Antibody (2B5)

Product Code. 15589012
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15589012 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15589012 Supplier Invitrogen Supplier No. MA521593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

REXO2 Monoclonal Antibody (2B5)

REXO2 Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen REXO2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene REXO2
Gene Accession No. Q9Y3B8
Gene Alias 1810038D15Rik; AW107347; C11orf61; CGI-114; Gm13138; novel KRAB box domain containing protein; oligoribonuclease, mitochondrial; OTTMUSG00000010655; reduced expression 2; REX2; REX2, RNA exonuclease 2 homolog; REX2, RNA exonuclease 2 homolog (S. cerevisiae); Rexo2; RFN; RNA exonuclease 2; RNA exonuclease 2 homolog; SFN; small fragment nuclease; SMFN
Gene Symbols REXO2
Host Species Mouse
Immunogen REXO2 (NP_056338, 101 a.a. ~ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25996
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.