missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RF1ML Monoclonal Antibody (OTI1B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525064
This item is not returnable.
View return policy
Description
RF1ML Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.
Specifications
| RF1ML | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UGC7 | |
| MTRF1L | |
| Full length human recombinant protein of MTRF1L produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1B3 | |
| Unconjugated | |
| MTRF1L | |
| 9130004K12Rik; HMRF1L; mitochondrial release factor 1 like; mitochondrial translational release factor 1 like; mitochondrial translational release factor 1-like; MRF1L; mtRF1a; MTRF1L; Peptide chain release factor 1-like, mitochondrial | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54516 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction