missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-961AF647
This item is not returnable.
View return policy
Description
RFC1 Polyclonal antibody specifically detects RFC1 in Human samples. It is validated for Western Blot
Specifications
| RFC1 | |
| Polyclonal | |
| Western Blot | |
| A1, A1 140 kDa subunit, Activator 1 140 kDa subunit, Activator 1 large subunit, Activator 1 subunit 1, MGC51786, MHC binding factor, beta, MHCBFB, PO-GA, RECC1, replication factor C (activator 1) 1 (145kD), replication factor C (activator 1) 1, 145kDa, Replication factor C 140 kDa subunit, Replication factor C large subunit, replication factor C subunit 1, replication factor C1, RFC, RF-C 140 kDa subunit, RFC140DNA-binding protein PO-GA | |
| The immunogen recognized by this antibody maps to a region between residue 1100 and the C-terminus (residue 1148) of human Replication Factor C subunit 1 using the numbering given in entry NP_002904.3 (GeneID 5981). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 5981 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction