missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC2 Antibody (OTI11C6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73856PCP
This item is not returnable.
View return policy
Description
RFC2 Monoclonal specifically detects RFC2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| RFC2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| RFC2 | |
| Full length recombinant protein of human RFC2 (NP_002905) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI11C6 | |
| Flow Cytometry | |
| A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD) | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, DNA Repair | |
| 5982.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction