Learn More
RFC2 Mouse anti-Human, Clone: OTI11C6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505002
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also called activator 1, is a protein complex consisting of five distinct subunits of 145, 40, 38, 37, and 36.5 kD. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP. Deletion of this gene has been associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| RFC2 | |
| Monoclonal | |
| Unconjugated | |
| RFC2 | |
| RFC40 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5982 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI11C6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P35250 | |
| RFC2 | |
| Full length human recombit protein of human RFC2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.