missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556169
This item is not returnable.
View return policy
Description
Immunogen sequence: PAPAFSKAPG SAGHYELPWV EKYRPVKLNE IVGNEDTVSR LEVFAREGNV PNIIIAGPPG TGKTTSILCL ARALLGPALK DAMLELNASN D Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene encodes a member of the activator 1 small subunits family. The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). Replication factor C, also called activator 1, is a protein complex consisting of five distinct subunits. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP and may help promote cell survival. Disruption of this gene is associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene of this gene has been defined on chromosome 2.
Specifications
| RFC2 | |
| Polyclonal | |
| Unconjugated | |
| RFC2 | |
| 2610008M13Rik; 40kDa; A1 40 kDa subunit; Activator 1 40 kDa subunit; activator 1 subunit 2; Activator 1 subunit C2; AI326953; MGC3665; Recc2; replication factor C (activator 1) 2; replication factor C (activator 1) 2, 40kDa; replication factor C 40 kDa subunit; Replication factor C subunit 2; replication factor C, 40 kDa; RF-C 40 kDa subunit; RFC2; RFC40 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116468, 19718, 5982 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35250, Q641W4, Q9WUK4 | |
| RFC2 | |
| Recombinant protein corresponding to Human RFC2. Recombinant protein control fragment (Product #RP-92618). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction