missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RFC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583126
This item is not returnable.
View return policy
Description
Immunogen sequence: LSTVCKKEGL NLPSQLAHRL AEKSCRNLRK ALLMCEACRV QQYPFTADQE IPETDWEVYL RETANAIVSQ QTPQRLLEVR GRLYELLTHC IPPEIIMKGL LSELLHNCDG QLKGEVAQMA AYYEHRLQLG SKAIYHLEAF VA Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.
The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kD. This gene encodes the 38 kD subunit. This subunit is essential for the interaction between the 140 kD subunit and the core complex that consists of the 36, 37, and 40 kD subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| RFC3 | |
| Polyclonal | |
| Unconjugated | |
| RFC3 | |
| 2810416I22Rik; 38kDa; A1 38 kDa subunit; Activator 1 38 kDa subunit; activator 1 subunit 3; AU022547; DNA-binding; MGC5276; Polymorphism; Recc3; replication factor C (activator 1) 3; replication factor C (activator 1) 3, 38kDa; replication factor C 3; replication factor C 38 kDa subunit; Replication factor C subunit 3; replication factor C, 38 kDa; RF-C 38 kDa subunit; RFC, 38 kD subunit; RFC3; RFC38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P40938 | |
| RFC3 | |
| Recombinant protein corresponding to Human RFC3. Recombinant protein control fragment (Product #RP-88828). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction