missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC4 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-403AF405
This item is not returnable.
View return policy
Description
RFC4 Polyclonal antibody specifically detects RFC4 in Human samples. It is validated for Western Blot
Specifications
| RFC4 | |
| Polyclonal | |
| Western Blot | |
| A1, A1 37 kDa subunit, Activator 1 37 kDa subunit, Activator 1 subunit 4, MGC27291, replication factor C (activator 1) 4, 37kDa, Replication factor C 37 kDa subunit, replication factor C subunit 4, RFC 37 kDa subunit, RF-C 37 kDa subunit, RFC37replication factor C (activator 1) 4 (37kD) | |
| A 23 amino acid synthetic peptide, which represented a portion of the human Replication Factor C 4, 37 kDa subunit encoded within exon 2 (locusLink ID 5984). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 5984 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction