missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL1 Antibody (OTI1D3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73861APC
This item is not returnable.
View return policy
Description
RFPL1 Monoclonal specifically detects RFPL1 in Human samples. It is validated for Flow Cytometry.
Specifications
| RFPL1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| RFPL1 | |
| Full length recombinant protein of human RFPL1 (NP_066306) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| OTI1D3 | |
| Flow Cytometry | |
| MGC132428, ret finger protein-like 1, RFPL1L, RNF78RING finger protein 78 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 5988.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction