missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trypsin 2/PRSS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93609-0.02ml
This item is not returnable.
View return policy
Description
Trypsin 2/PRSS2 Polyclonal antibody specifically detects Trypsin 2/PRSS2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Trypsin 2/PRSS2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Anionic trypsinogen, EC 3.4.21, EC 3.4.21.4, MGC111183, MGC120174, MGC120175, protease serine 2 preproprotein, protease, serine, 2 (trypsin 2), protease, serine, 2, preproprotein, Serine protease 2, TRY2TRY8, TRYP2, trypsin 2, Trypsin II, trypsin-2, trypsinogen 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-247 of human PRSS2 (NP_002761.1). IVGGYICEENSVPYQVSLNSGYHFCGGSLISEQWVVSAGHCYKSRIQVRLGEHNIEVLEGNEQFINAAKIIRHPKYNSRTLDNDILLIKLSSPAVINSRVSAISLPTAPPAAGTESLISGWGNTLSSGADYPDELQCLDAPVLSQAECEASYPGKITNNMFCVGFLEGGKDSCQGDSGGPVVSNGELQGIVSWGYGCAQKNRPGVYTKVYNYVDWIKDTIAANS | |
| 0.02 mL | |
| Cell Biology | |
| 5645 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction