missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL3 Antibody (OTI2G4), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73862PEATT594
This item is not returnable.
View return policy
Description
RFPL3 Monoclonal specifically detects RFPL3 in Human samples. It is validated for Flow Cytometry.
Specifications
| RFPL3 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| RFPL3 | |
| Full length recombinant protein of human RFPL3 (NP_006595) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| OTI2G4 | |
| Flow Cytometry | |
| ret finger protein-like 3 | |
| Mouse | |
| Affinity Purified | |
| Zinc Finger | |
| 10738.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction