missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL4A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | RFPL4A |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RFPL4A Polyclonal antibody specifically detects RFPL4A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RFPL4A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 342931 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ret finger protein-like 4A, RFPL4, RING finger protein 210, RNF210ret finger protein-like 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RFPL4 (NP_001138486.1). MAEHFKQIIRCPVCLKDLEEAVQLKCGYACCLQCLNSLQKEPDGEGLLCRFCSVVSQKDDIKPKYKLRALVSIIKELEPKLKSVLTMNPRMRKFQVDMTF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title