missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52653UV
This item is not returnable.
View return policy
Description
RFX1 Polyclonal antibody specifically detects RFX1 in Human samples. It is validated for Western Blot
Specifications
| RFX1 | |
| Polyclonal | |
| Western Blot | |
| EFC, EF-C, Enhancer factor C, MHC class II regulatory factor RFX, MHC class II regulatory factor RFX1, Regulatory factor X 1, regulatory factor X, 1 (influences HLA class II expression), RFX, trans-acting regulatory factor 1, Transcription factor RFX1 | |
| The immunogen for this product maps to a region between residue 200 and 250 of human Regulatory Factor X, 1 using the numbering given in entry NP_002909.4 (GeneID 5989). | |
| 0.1 mL | |
| Immunology | |
| 5989 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction