missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX5 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-195FR
This item is not returnable.
View return policy
Description
RFX5 Polyclonal antibody specifically detects RFX5 in Human samples. It is validated for Western Blot
Specifications
| RFX5 | |
| Polyclonal | |
| Western Blot | |
| DNA-binding protein RFX5, Regulatory factor X 5, regulatory factor X, 5 (influences HLA class II expression) | |
| Rabbits were immunized with a synthetic peptide, which represented a portion of human Regulatory Factor X 5 encoded within exon 5 (LocusLink ID 5993), conjugated to KLH. After multiple immunizations in Freund's adjuvant, serum was collected. Antibodies were affinity purified using the peptide immobilized on solid support. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 5993 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction