missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFX8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15908994
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15908994 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15908994 Supplier Invitrogen™ Supplier No. PA566278

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LLQISKLKEVT LFVKRLRRKT YLSNMAKTMR MVLKSKRRVS VLKSDLQAII NQGTLATSKK ALASDRSGAD ELENNPEMKC L Highest antigen sequence identity to the following orthologs - mouse 70%, rat 71%.

RFX8 may be a transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFX8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RFX8
Gene Accession No. Q6ZV50
Gene Alias DNA-binding protein RFX8; Regulatory factor X 8; regulatory factor X, 8; RFX family member 8, lacking RFX DNA binding domain; RFX gene family member 8, lacking RFX DNA binding domain; RFX8
Gene Symbols RFX8
Host Species Rabbit
Immunogen Recombinant Human RFX8. Recombinant protein control fragment (Product #RP-105138).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 731220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.