missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFXANK Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97640AF594
This item is not returnable.
View return policy
Description
RFXANK Polyclonal specifically detects RFXANK in Human samples. It is validated for Western Blot.
Specifications
| RFXANK | |
| Polyclonal | |
| Western Blot | |
| ANKRA1Regulatory factor X subunit B, ankyrin repeat-containing regulatory factor X-associated protein, BLS, DNA-binding protein RFXANK, F14150_1, MGC138628, regulatory factor X-associated ankyrin-containing proteinAnkyrin repeat family A protein 1, RFXB, RFX-BRFX-Bdelta4 | |
| Produced in rabbits immunized with E. coli-derived Human RFXANK fragment. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 8625 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction