missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFXANK Antibody (OTI3B4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73868H
This item is not returnable.
View return policy
Description
RFXANK Monoclonal specifically detects RFXANK in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RFXANK | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RFXANK | |
| Full length recombinant protein of human RFXANK (NP_604389) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI3B4 | |
| Western Blot | |
| ANKRA1Regulatory factor X subunit B, ankyrin repeat-containing regulatory factor X-associated protein, BLS, DNA-binding protein RFXANK, F14150_1, MGC138628, regulatory factor X-associated ankyrin-containing proteinAnkyrin repeat family A protein 1, RFXB, RFX-BRFX-Bdelta4 | |
| Mouse | |
| Affinity Purified | |
| Adaptive Immunity, Immunology | |
| 8625.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction