Learn More
RFXANK Mouse anti-Human, Clone: OTI3F5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504270
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Major histocompatibility (MHC) class II molecules are transmembrane proteins that have a central role in development and control of the immune system. The protein encoded by this gene, along with regulatory factor X-associated protein and regulatory factor-5, forms a complex that binds to the X box motif of certain MHC class II gene promoters and activates their transcription. Once bound to the promoter, this complex associates with the non-DNA-binding factor MHC class II transactivator, which controls the cell type specificity and inducibility of MHC class II gene expression. This protein contains ankyrin repeats involved in protein-protein interactions. Mutations in this gene have been linked to bare lymphocyte syndrome type II, complementation group B. Two transcript variants encoding different isoforms have been described for this gene, with only one isoform showing activation activity.Specifications
| RFXANK | |
| Monoclonal | |
| Unconjugated | |
| RFXANK | |
| ANKRA1, BLS, F14150_1, RFX-B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8625 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O14593 | |
| RFXANK | |
| Full length human recombit protein of human RFXANK produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.