missing translation for 'onlineSavingsMsg'
Learn More

RG13022, MedChemExpress

Product Code. 16409896
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409896

Brand: Medchem Express HY101429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG13022 is a tyrosine kinase inhibitor; inhibits the autophosphorylation reaction of the EGF receptor with an IC50 of 4 μM.

RG13022 is a tyrosine kinase inhibitor

Specifications

Chemical Name or Material RG13022
CAS 136831-48-6
Color White
Purity Grade Notes Research
Molecular Formula C16H14N2O2
Quantity 50 mg
Synonym Tyrphostin RG13022
Solubility Information DMSO : 100 mg/mL (375.53 mM; Need ultrasonic)
SMILES N#C/C(C1=CC=CN=C1)=C/C2=CC=C(OC)C(OC)=C2
Molecular Weight (g/mol) 266.29
Formula Weight 266.29
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.