Learn More
RG14620, MedChemExpress
Product Code. 16479886
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16479886
Brand: Medchem Express HY101426
MedChemExpress RG14620 is an EGFR inhibitor with an IC50 of 3 μM.
RG14620 is an EGFR inhibitor with an IC50 of 3 μM.
Specifications
Chemical Name or Material RG14620
Physical Form Solid
CAS 136831-49-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H8Cl2N2
For Use With (Application) Cancer-Kinase/protease
Synonym Tyrphostin RG14620
Solubility Information DMSO : 33.33 mg/mL (121.14 mM; Need ultrasonic)
SMILES N#C/C(C1=CC=CN=C1)=C/C2=CC(Cl)=CC(Cl)=C2
Molecular Weight (g/mol) 275.13
Formula Weight 275.13
Percent Purity 99.85%
Grade Research
Show More Show Less
Research purposes only