missing translation for 'onlineSavingsMsg'
Learn More

RG7713, MedChemExpress

Product Code. 16471319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471319 1 mg 1mg
16471637 50 mg 50mg
16448177 5 mg 5mg
16436056 100 mg 100mg
16405468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471319 Supplier Medchem Express Supplier No. HY12981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a).

RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a).

Specifications

Chemical Name or Material RG7713
CAS 920022-47-5
Color White
Purity Grade Notes Research
Molecular Formula C25H28ClN3O2
Quantity 1 mg
Synonym RO5028442
Solubility Information DMSO : 20 mg/mL (45.67 mM; Need ultrasonic)
SMILES O=C(C1=CN(CCN(C)C)C2=C1C=CC(Cl)=C2)N3CCC4(C(C=CC=C5)=C5CO4)CC3
Molecular Weight (g/mol) 437.96
Formula Weight 437.96
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.