Learn More
Invitrogen™ RGD1308134 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA569934
Product Code. 15848343
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%.
RGD1308134 gene ontology annotations related to this gene include DNA binding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGD1308134
Polyclonal
Unconjugated
RGD1308134
chromatin complexes subunit BAP18; RGD1308134; similar to RIKEN cDNA 1110020A23
Rabbit
Affinity chromatography
RUO
287452
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
RGD1308134
synthetic peptide directed towards the N-terminal region of rat RGD1308134.