missing translation for 'onlineSavingsMsg'
Learn More

RGFP966, MedChemExpress

Product Code. 16408547
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16408547

Brand: Medchem Express HY13909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. RGFP966 can penetrate the blood brain barrier (BBB).

RGFP966 is a highly selective HDAC3 inhibitor with an IC50 of 80 nM and shows no inhibition to other HDACs at concentrations up to 15 μM. RGFP966 can penetrate the blood brain barrier (BBB).

Specifications

Chemical Name or Material RGFP966
CAS 1357389-11-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19FN4O
Quantity 5 mg
Solubility Information DMF : 50 mg/mL (137.97 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (137.97 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(F)C=C1N)/C=C/C2=CN(C/C=C/C3=CC=CC=C3)N=C2.[E]
Molecular Weight (g/mol) 362.4
Formula Weight 362.4
Percent Purity 99.13%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.