missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110289

Product Code. 17188213

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LKTQQRAKII EKWINIAHEC RLLKNFSSLR AIVSALQSNS IYRLKKTWAA VPRDRMLMFE ELSDIFSDHN NHLTSRELLM KEGTSKFANL DSSVKENQKR T.

Ral GDS (Ral guanine nucleotide dissociation stimulator) is a guanine nucleotide exchange factor (GEF) that activates Ral and is implicated in oncogenic Ras-induced cell transformation. RGL1 (ral guanine nucleotide dissociation stimulator-like 1), also known as RGL or RalGDS-like 1, is a 768 amino acid protein that is a putative GEF. Strongly expressed in brain, heart, spleen, kidney and testis, RGL1 is a downstream effector protein that is involved in Ras and Ral signaling pathways. RGL1 contains an N-terminal Ras-GEF domain and a C-terminal Ras-interacting domain that interacts with the GTP-bound form of Ras through its effector loop. Due to its similarity to Ral GDS, RGL1 may be implicated in carcinogenesis. Two isoforms exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGL1
Polyclonal
Unconjugated
RGL1
KIAA0959; mKIAA0959; ral guanine nucleotide dissociation stimulator like 1; ral guanine nucleotide dissociation stimulator,-like 1; ral guanine nucleotide dissociation stimulator-like 1; RalGDS-like 1; RGL; Rgl1
Rabbit
Affinity chromatography
RUO
19731, 23179, 289080
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.11 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q60695, Q9NZL6
RGL1
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human RGL1 (NP_0012846001).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.