Learn More
RGM-A Antibody (elezanumab) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28156JF585
Product Code. 30178061
  • € 431.00 / 0.10mL
Description
RGM-A Monoclonal antibody specifically detects RGM-A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGM-A
Monoclonal
Janelia Fluor 585
RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A
RGMA
0.1 mL
Neuroscience, Stem Cells
56963
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
elezanumab
50mM Sodium Borate
Human
Protein A purified