Learn More
Invitrogen™ RGMA Recombinant Rabbit Monoclonal Antibody (2S1B4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542630
Description
Positive test controls include: Mouse heart, Rat skeletal muscle,Rat heart. The target is usually found in the following locations: Cell membrane, GPI-anchor, Lipid-anchor. Immunogen sequence: TAPETFPYET AVAKCKEKLP VEDLYYQACV FDLLTTGDVN FTLAAYYALE DVKMLHSNKD KLHLYERTRD LPGRAAAGLP LAPRPLLGAL VPLLALLPVF C.
Member of the repulsive guidance molecule (RGM) family that performs several functions in the developing and adult nervous system. Regulates cephalic neural tube closure, inhibits neurite outgrowth and cortical neuron branching, and the formation of mature synapses. Binding to its receptor NEO1/neogenin induces activation of RHOA-ROCK1/Rho-kinase signaling pathway through UNC5B-ARHGEF12/LARG-PTK2/FAK1 cascade, leading to collapse of the neuronal growth cone and neurite outgrowth inhibition. Furthermore, RGMA binding to NEO1/neogenin leads to HRAS inactivation by influencing HRAS-PTK2/FAK1-AKT1 pathway. It also functions as a bone morphogenetic protein (BMP) coreceptor that may signal through SMAD1, SMAD5, and SMAD8.
Specifications
| RGMA | |
| Recombinant Monoclonal | |
| 0.67 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PCX7 | |
| RGMA | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human RGMA (Q96B86). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2S1B4 | |
| Unconjugated | |
| RGMA | |
| BC059072; C230063O06; repulsive guidance molecule; repulsive guidance molecule A; repulsive guidance molecule family member a; RGM; RGM domain family member A; RGM domain family, member A; RGMA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 244058, 308739 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.