Learn More
Invitrogen™ RGMB Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599857
Product Code. 16634105
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of total RGMB.
Repulsive Guidance Molecule B (RGMB) is primarily expressed in the central nervous system. RGM A and B both appear at every level of the developing neural axis, where they colocalize to a large extent in the mantle layer, although only RGMA appears in the neuroepithelium, and only RGMB in the peripheral nervous system. During development, both RGMA and B appear also in lung and in limb cartilage, while RGMB has additional expression domains in pancreas.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RGMB
Polyclonal
Unconjugated
RGMB
1110059F19Rik; DRAGON; DRG11-responsive axonal guidance and outgrowth of neurite; repulsive guidance molecule B; repulsive guidance molecule family member B; RGM domain family member B; RGM domain family, member B; RGMB
Rabbit
Affinity chromatography
RUO
285704
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6NW40
RGMB
A synthesized peptide derived from human RGMB(Accession Q6NW40), corresponding to amino acid residues L110-Y160.