missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGNEF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22282
This item is not returnable.
View return policy
Description
RGNEF Polyclonal specifically detects RGNEF in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RGNEF | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| 190 kDa guanine nucleotide exchange factor, p190RHOGEF, Rho-guanine nucleotide exchange factor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64283 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_001171164.1 | |
| ARHGEF28 | |
| The immunogen this antibody was made to, maps to a region between residue 1655 and 1705 of human 190 kDa, Rho-Guanine Nucleotide Exchange Factor using the numbering given in entry NP_001171164.1 (GeneID 64283). | |
| 100 μL | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Crab-eating macaque and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction