missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGPD Pan Polyclonal Antibody

Description
RGPD Pan Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RGPD Pan |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RGPD2 |
| Gene Accession No. | A6NKT7, O14715, P0DJD1, Q7Z3J3, Q99666 |
| Gene Alias | BS63; BS-63; epididymis luminal protein 161; HEL161; nucleoporin; NUP358; RAN binding protein 2-like 1; Ran binding protein 2-like 2; ran-binding protein 2-like 1; ran-binding protein 2-like 1/2; Ran-binding protein 2-like 2; ran-binding protein 2-like 3; RanBP2alpha; RANBP2L1; RANBP2L2; RANBP2L3; ranBP2-like 1; ranBP2-like 1/2; RanBP2-like 2; ranBP2-like 3; RANBP2-like and GRIP domain containing 2; RANBP2-like and GRIP domain containing 3; RANBP2-like and GRIP domain containing 4; RANBP2-like and GRIP domain containing 5; RANBP2-like and GRIP domain containing 8; RANBP2-like and GRIP domain-containing protein 2; RanBP2-like and GRIP domain-containing protein 3; ranBP2-like and GRIP domain-containing protein 4; RANBP2-like and GRIP domain-containing protein 5; RANBP2-like and GRIP domain-containing protein 5/6; RANBP2-like and GRIP domain-containing protein 8; RGP2; RGP3; RGP4; RGP5; RGP6; RGP7; RGP8; RGPD2; RGPD3; RGPD4; RGPD5; RGPD6; RGPD7; RGPD8; Sperm membrane protein BS-63 |
| Gene Symbols | RGPD2, RGPD3, RGPD4, RGPD5, RGPD8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?