missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562456
This item is not returnable.
View return policy
Description
Immunogen sequence: SETSSVAQSG SESKVEPKKC ELSKNSDIEQ SSDSKVK Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
RanBP2 interacts with the small GTP-binding protein Ran and forms a large multiprotein complex involved in many biological processes such as mRNA processing, nuclear transport, cell cycle control and postmitotic nuclear reassembly. The protein is identified by western blotting as an apparent molecular mass of 360 kD. It is involved in nucleation of microtubule and control of cell growth by interacting with many protein factors.
Specifications
| RANBP2 | |
| Polyclonal | |
| Unconjugated | |
| RANBP2 | |
| 358 kDa nucleoporin; A430087B05Rik; acute necrotizing encephalopathy 1 (autosomal dominant); ADANE; AI256741; ANE1; E3 SUMO-protein ligase RanBP2; E3 SUMO-protein transferase RanBP2; IIAE3; Nuclear pore complex protein Nup358; nucleoporin 358; nucleoporin Nup358; NUP358; P270; RAN binding protein 2; ran-binding protein 2; RANBP2; RBP2; retina-specific cyclophilin; RGD1560047; spliced variant with Ran-binding and cyclophilin domains; transformation-related protein 2; TRP1; TRP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5903 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49792 | |
| RANBP2 | |
| Recombinant protein corresponding to Human RGPD1. Recombinant protein control fragment (Product #RP-104465). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction