missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS10 Monoclonal Antibody (OTI8H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812338S
This item is not returnable.
View return policy
Description
RGS10 Monoclonal antibody specifically detects RGS10 in Human samples. It is validated for Western BlotSpecifications
| RGS10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92993 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10524 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8H4 | |
| Unconjugated | |
| KAT5 | |
| 2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10) | |
| Human recombinant protein fragment corresponding to amino acids 227-504 of human KAT5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction