missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560210
This item is not returnable.
View return policy
Description
Immunogen sequence: GDRLVIACQE QTYWLVNRPP PGAPDVLEQG PGRGSCAASR VLMTKSADFH KREIEYFRKA LGRTRVKSSV CLEAYLSFCG QRGPHDPL Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 38%.
The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
| RGS11 | |
| Polyclonal | |
| Unconjugated | |
| RGS11 | |
| AC004754.2; C78048; regulator of G-protein signaling 11; regulator of G-protein signalling 11; RGS11; RS11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94810 | |
| RGS11 | |
| Recombinant protein corresponding to Human RGS11. Recombinant protein control fragment (Product #RP-98614). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction