missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AU5 Epitope Tag Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-458F
This item is not returnable.
View return policy
Description
AU5 Epitope Tag Polyclonal antibody specifically detects AU5 Epitope Tag in Epitope Tag samples. It is validated for ELISA, Immunofluorescence
Specifications
| AU5 Epitope Tag | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Purified | |
| Epitope Tag |
| ELISA, Immunofluorescence | |
| FITC | |
| AU5 epitope tag, TDFYLK epitope tag, TDFYLK tag | |
| Rabbits were immunized with AU5 (TDFYLK) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
| 0.1 mL | |
| Epitope Tags | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction