missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101834
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RGS12.
This gene encodes a member of the 'regulator of G protein signaling' (RGS) gene family. The encoded protein may function as a guanosine triphosphatase (GTPase)-activating protein as well as a transcriptional repressor. This protein may play a role in tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their biological nature has not been determined.
Specifications
| RGS12 | |
| Polyclonal | |
| Unconjugated | |
| RGS12 | |
| 1200016K18Rik; 4632412M04Rik; AI481290; DKFZp761K1617; DKFZp761K1817; E130309H11; regulator of G-protein signaling 12; regulator of G-protein signalling 12; Rgs12; RP11-529E10.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54292, 6002, 71729 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08774, O14924, Q8CGE9 | |
| RGS12 | |
| A synthesized peptide derived from human RGS12(Accession O14924), corresponding to amino acid residues E140-N190. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction