missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20147
This item is not returnable.
View return policy
Description
RGS14 Polyclonal specifically detects RGS14 in Human samples. It is validated for Western Blot.
Specifications
| RGS14 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| regulator of G-protein signaling 14, regulator of G-protein signalling 14 | |
| Rabbit | |
| 61 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 10636 | |
| Human | |
| IgG |
| Western Blot | |
| 0.79mg/mL | |
| Western Blot 1:500-1:3000 | |
| O43566 | |
| RGS14 | |
| Recombinant protein encompassing a sequence within the center region of human RGS14. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (84%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction