missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS14 Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | RGS14 |
|---|---|
| Clone | 1C1B2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16879995
|
Proteintech
67394-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16869995
|
Proteintech
67394-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RGS14 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RGS14. This antibody recognizes human, mouse, rat antigen. The RGS14 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| RGS14 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 10636 | |
| Primary |
| 1C1B2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title