missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS16 Antibody (OTI2C9), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73884FR
This item is not returnable.
View return policy
Description
RGS16 Monoclonal specifically detects RGS16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RGS16 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RGS16 | |
| Full length recombinant protein of human RGS16 (NP_002919) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2C9 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| A28-RGS14, A28-RGS14P, hRGS-r, regulator of G-protein signaling 16, regulator of G-protein signalling 16, Retinally abundant regulator of G-protein signaling, Retinal-specific RGS, RGSR, RGS-R | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 6004.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction