missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584143
This item is not returnable.
View return policy
Description
Immunogen sequence: MCRTLAAFPT TCLERAKEFK TRLGIFLHKS ELGCDTGSTG KFEWGSKHSK ENRNFSED Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 83%.
The protein encoded by this gene belongs to the 'regulator of G protein signaling' family. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits. It also may play a role in regulating the kinetics of signaling in the phototransduction cascade.
Specifications
| RGS16 | |
| Polyclonal | |
| Unconjugated | |
| RGS16 | |
| A28-RGS14; A28-RGS14P; hRGS-r; regulator of G-protein signaling 16; regulator of G-protein signalling 16; Retinally abundant regulator of G-protein signaling; retinal-specific RGS; Rgs14; RGS16; RGSR; RGS-R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6004 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15492 | |
| RGS16 | |
| Recombinant protein corresponding to Human RGS16. Recombinant protein control fragment (Product #RP-101353). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction