missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS17 Monoclonal Antibody (OTI6G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811296S
This item is not returnable.
View return policy
Description
RGS17 Monoclonal antibody specifically detects RGS17 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RGS17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11217 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G3 | |
| Unconjugated | |
| PYGM | |
| 6430507P11Rik; hRGS17; regulator of G-protein signaling 17; regulator of G-protein signaling Z2; regulator of Gz-selective protein signaling 2; Rgs17; RGS-17; Rgsz2 | |
| Human recombinant protein fragment corresponding to amino acids 698-842 of human PYGM produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction