Learn More
RGS17 Monoclonal Antibody (OTI8G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811301
This item is not returnable.
View return policy
Description
This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.Specifications
| RGS17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UGC6 | |
| RGS17 | |
| Full length human recombit protein of human RGS17 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8G4 | |
| Unconjugated | |
| RGS17 | |
| RGS-17, RGSZ2, hRGS17 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |