missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62667-25ul
This item is not returnable.
View return policy
Description
RGS20 Polyclonal antibody specifically detects RGS20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RGS20 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| G(z)GAP, Gz-GAP, Gz-selective GTPase-activating protein, regulator of G-protein signaling 20, Regulator of G-protein signaling Z1, regulator of G-protein signalling 20, Regulator of Gz-selective protein signaling 1, RGSZ1g(z)GAP, ZGAP1gz-GAP | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MPQLSQDNQECLQKHFSRPSIWTQFLPLFRAQRYNTDIHQITENEGDLRAVP | |
| 25 μL | |
| Signal Transduction | |
| 8601 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction