missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody (OTI1C1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73887F
This item is not returnable.
View return policy
Description
RGS5 Monoclonal specifically detects RGS5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| RGS5 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| RGS5 | |
| Full length recombinant protein of human RGS5 (NP_003608) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI1C1 | |
| Flow Cytometry, Immunohistochemistry | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 8490.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction