Learn More
RGS5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | RGS5 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069425
|
Proteintech
CL488-11590-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18005205
|
Proteintech
CL488-11590-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RGS5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RGS5. This antibody recognizes human, mouse, rat antigen. The RGS5 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| RGS5 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, RGS5 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 8490 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.