missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06581-50ul
This item is not returnable.
View return policy
Description
RGS6 Polyclonal antibody specifically detects RGS6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RGS6 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| DKFZp313G1241, FLJ43552, G protein signaling 6 regulator, GAP, GTPase activating protein, H_DJ0283M22.1, H_DJ1108A12.1, MGC142132, regulator of G protein signaling 6, regulator of G protein signalling 6, regulator of G-protein signaling 6, regulator of G-protein signalling 6, S914, WUGSC:H_DJ0283M22.1, WUGSC:H_DJ1108A12.1 | |
| Produced in rabbits immunized with E. coli-derived Human RGS6 fragment. | |
| 50 μg | |
| Signal Transduction | |
| 9628 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction