missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94343-0.02ml
This item is not returnable.
View return policy
Description
RGS8 Polyclonal antibody specifically detects RGS8 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RGS8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC119067, MGC119068, MGC119069, regulator of G-protein signaling 8, regulator of G-protein signalling 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human RGS8 (NP_001095920.1). MAALLMPRRNKGMRTRLGCLSHKSDSCSDFTAILPDKPNRALKRLSTEEATRWADSFDVLLSHKYGVAAFRAFLKTEFSEENLEFWLACEEFKKTRSTAKLVSKAHRIFEEFVDVQAPREVNIDFQTREATRKNLQEPSLTCFDQAQGKVHSLMEKDSYPRFLRSKMYLDLLSQSQRRLS | |
| 0.02 mL | |
| Signal Transduction | |
| 85397 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction