missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531105
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (98%), Rat (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
A novel human RGS8 gene (homologue of the rat RGS8 gene) is expressed in prostate tissues and represents a positional candidate for the HPC1 locus, which is involved in hereditary prostate cancer and hyperparathyroidism-jaw tumor syndrome.
Specifications
| RGS8 | |
| Polyclonal | |
| Unconjugated | |
| RGS8 | |
| 6530413N01Rik; Regulator of G-protein signaling 8; regulator of G-protein signalling 8; Rgs8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 85397 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P57771 | |
| RGS8 | |
| Recombinant fragment corresponding to a region within amino acids 1 and 180 of Human RGS8. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction